Please login to your online account to display your discounted pricing

Potassium Iodate-Iodide, 0.0250N (N/40), Aqueous Solution, Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR628200020F

 View more versions of this product

Catalog No. R628200020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 20L
Chemical Name or Material Potassium Iodate-Iodide
CAS 7758-05-6
CAS 7681-11-0
CAS 7732-18-5
Name Note 0.0250N (N/40)
Assay Percent Range 0.02500 ± 0.00001 N at 20°C
Odor Odorless
Packaging Cubitainer
Solubility Solubility in Water: Infinite
CAS 1310-73-2
Solvent or Matrix Aqueous Solution

Specifications

Chemical Name or Material Potassium Iodate-Iodide
Name Note 0.0250N (N/40)
CAS 7758-05-6
CAS 7681-11-0
CAS 1310-73-2
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 0.02500 ± 0.00001 N at 20°C