Please login to your online account to display your discounted pricing

Potassium Iodate-Iodide, 0.0250N (N/40), Aqueous Solution, Ricca Chemical

$147.21 - $147.21

Chemical Identifiers

Chemical Name or Material Potassium Iodate-Iodide
Name Note 0.0250N (N/40)
CAS 7758-05-6
CAS 7681-11-0
CAS 1310-73-2
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 0.02500 ± 0.00001 N at 20°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R628200020F

 
ricca chemical company
RIR628200020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R62820004C

 
ricca chemical company
RIR62820004C
Amber glass 4L Each for $147.21
Description & Specifications

Specifications

Chemical Name or Material Potassium Iodate-Iodide
Name Note 0.0250N (N/40)
CAS 7758-05-6
CAS 7681-11-0
CAS 1310-73-2
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 0.02500 ± 0.00001 N at 20°C
Odor Odorless
Packaging Cubitainer
Solubility Solubility in Water: Infinite
Solvent or Matrix Aqueous Solution
Color Clear colorless
Quantity 20L