Please login to your online account to display your discounted pricing

Potassium Iodate, For Chlorine, Certified, 0.00564N ±0.00010N (0.00094M), LabChem™

$100.94 - $100.94

Chemical Identifiers

Chemical Name or Material Potassium Iodate
CAS 7732-18-5
CAS Max % 99.98
CAS 7758-05-6
CAS Max % 0.02
Name Note For Chlorine
Purity Grade Certified Grade
Linear Formula KIO3
Formula Weight 214
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC196002

 
lab chem
LC196002
1L Each for $100.94
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 214
Quantity 1L
Chemical Name or Material Potassium Iodate
Density 1g/mL
Concentration 0.00564N ±0.00010N (0.00094M)
CAS 7758-05-6
Color Clear, colorless
CAS 7732-18-5
Solubility Soluble in water
Packaging Poly Bottle
CAS Max % 99.98
Decomposition Information Iodine vapor
Storage Information Room Temperature
Identification Passes Test
CAS Max % 0.02
Traceability to NIST Traceable to NIST
Linear Formula KIO3
Name Note For Chlorine
Physical Form Liquid
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Specific Gravity 1g/mL
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.