Please login to your online account to display your discounted pricing

Potassium Iodate, Certified, 0.1250N ±0.0005N (0.0208M), LabChem™

$66.69 - $66.69

Chemical Identifiers

Chemical Name or Material Potassium Iodate
CAS 7732-18-5
CAS Max % 99.55
CAS 7758-05-6
CAS Max % 0.45
Purity Grade Certified Grade
Linear Formula KIO3
Formula Weight 214
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC196402

 
lab chem
LC196402
1L Each for $66.69
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 214
Chemical Name or Material Potassium Iodate
Identification Passes Test
Traceability to NIST Traceable to NIST
Packaging Poly Bottle
Physical Form Liquid
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Linear Formula KIO3
CAS 7732-18-5
Decomposition Information Iodine vapor
Color Clear, colorless
Quantity 1L
CAS Max % 99.55
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 7758-05-6
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.45
Concentration 0.1250N ±0.0005N (0.0208M)
Density 1g/mL
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.