Please login to your online account to display your discounted pricing

Potassium Iodate, Certified, 0.0250N ±0.0002N (0.0042M), LabChem™

$102.00 - $102.00

Chemical Identifiers

Chemical Name or Material Potassium Iodate
CAS 7732-18-5
CAS Max % 99.91
CAS 7758-05-6
CAS Max % 0.09
Purity Grade Certified Grade
Linear Formula KIO3
Formula Weight 214
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC196102

 
lab chem
LC196102
1L Each for $102.00
Description & Specifications

Specifications

Formula Weight 214
Purity Grade Certified Grade
Quantity 1L
CAS Max % 99.91
Linear Formula KIO3
Identification Passes Test
Concentration 0.0250N ±0.0002N (0.0042M)
CAS Max % 0.09
Chemical Name or Material Potassium Iodate
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Decomposition Information Iodine vapor
Color Clear, colorless
Specific Gravity 1g/mL
CAS 7758-05-6
Packaging Poly Bottle
Density 1g/mL
Solubility Soluble in water
Physical Form Liquid
Storage Information Room Temperature
CAS 7732-18-5
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.