Please login to your online account to display your discounted pricing

Potassium Iodate, ACS Grade, 99.4 to 100.4%, LabChem™

$145.20 - $145.20

Chemical Identifiers

Chemical Name or Material Potassium Iodate
CAS 7758-05-6
CAS Max % 100
Purity Grade ACS Grade
Linear Formula KIO3
Formula Weight 214
UN Number UN1479
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC195909

 
lab chem
LC195909
100g Each for $145.20
Description & Specifications

Specifications

Formula Weight 214
Decomposition Information Iodine vapor
Assay 99.4 to 100.4%
Specific Gravity 3930kg/m3
CAS Max % 100
Solubility Soluble in water
Purity Grade ACS Grade
Health Hazard 2 GHS H Statement
May intensify fire; oxidizer.
Chemical Name or Material Potassium Iodate
CAS 7758-05-6
Melting Point 560°C
Color White
Density 3,930kg/m3
Linear Formula KIO3
Physical Form Powder
Health Hazard 3 GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Wear protective gloves, protective clothing, eye protection, face protection.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Dispose of contents/container to comply with local, state and federal regulations.
EINECS Number 231-831-9
Packaging Poly Bottle
UN Number UN1479
Storage Information Room Temperature
Quantity 100g
Health Hazard 1 Warning
Identification Passes Test
Safety and Handling

Warning

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Wear protective gloves, protective clothing, eye protection, face protection.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May intensify fire; oxidizer.