missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium ICP Standard, 1000 ppm K in 3% HNO3, Ricca Chemical
$278.81 - $632.38
Chemical Identifiers
CAS | 7757-79-1 |
---|---|
Molecular Formula | KNO3 |
Molecular Weight (g/mol) | 101.10 |
MDL Number | MFCD00011409 |
InChI Key | FGIUAXJPYTZDNR-UHFFFAOYSA-N |
PubChem CID | 24434 |
ChEBI | CHEBI:63043 |
IUPAC Name | potassium nitrate |
SMILES | [K+].[O-][N+]([O-])=O |
Chemical Identifiers
7757-79-1 | |
101.10 | |
FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
CHEBI:63043 | |
[K+].[O-][N+]([O-])=O |
KNO3 | |
MFCD00011409 | |
24434 | |
potassium nitrate |
Specifications
7757-79-1,7697-37-2,7732-18-5 | |
100 mL | |
97.0,4.39,0.25 | |
KNO3 | |
FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
potassium nitrate | |
24434 | |
1mg K (1, 000ppm K)KNO3 in 3% HNO3 | |
Potassium ICP Standard |
Natural LDPE Bottle | |
94.0,4.22,0.24 | |
Colorless | |
MFCD00011409 | |
[K+].[O-][N+]([O-])=O | |
101.10 | |
CHEBI:63043 | |
Liquid |
Safety and Handling
ShelfLife : 18 months