Please login to your online account to display your discounted pricing

Potassium Hydroxide StandAR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Potassium Hydroxide
Name Note (w/v) Aqueous
CAS 1310-58-3
Assay Percent Range 10%
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MKH320100

 
avantor performance
H32001
Poly 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Potassium Hydroxide
Name Note (w/v) Aqueous
Assay Percent Range 10%
CAS 1310-58-3
Packaging Poly
Quantity 100mL