missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Honeywell Fluka™ Potassium hydroxide solution, Volumetric, 0.1 M KOH in ethanol (0.1N), Honeywell Fluka™
Volumetric, 0.1 M KOH in ethanol (0.1N)
Manufacturer: Honeywell Fluka™ 351276X1L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Potassium hydroxide solution | |
≥70.0000%,≥10.0000% | |
7732-18-5,108-88-3,1310-58-3 | |
KOH | |
Volumetric | |
14°C |
Volumetric, 0.1 M KOH in ethanol (0.1N) | |
<90.0000%,<20.0000% | |
0.1 M KOH in ethanol (0.1N) | |
alcohol, algrain, alkohol, anhydrol, ethyl alcohol, ethyl hydrate, ethyl hydroxide, grain alcohol, methylcarbinol, tecsol | |
Glass Bottle | |
6 x 1L |