Please login to your online account to display your discounted pricing

Potassium Hydroxide Solution AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Potassium Hydroxide Solution
Assay Percent Range 45%
Molecular Formula KOH
Formula Weight 56.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK66714

 
avantor performance
667104
Poly 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK6671500

 
avantor performance
667102
Poly 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 56.11
Chemical Name or Material Potassium Hydroxide Solution
Assay Percent Range 45%
Molecular Formula KOH
CAS 1310-58-3
Packaging Poly
Quantity 4L