Please login to your online account to display your discounted pricing

Potassium Hydroxide Solution, 45%(w/w), BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$498.05 - $1,349.61

Chemical Identifiers

Chemical Name or Material Potassium Hydroxide Solution
CAS 1310-58-3
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula KOH
Formula Weight 56.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003768

 
avantor performance
314303
Poly Bottle 4L Each for $498.05

02003769

 
avantor performance
314307
Jerrican (Poly Pail) 19L Each for $1,349.61
Description & Specifications

Specifications

Formula Weight 56.11
Quantity 4L
Chemical Name or Material Potassium Hydroxide Solution
Purity Grade BAKER ANALYZED™ Reagent
Assay 45% (w/w)
Specific Gravity 1L = 1.45kg
Molecular Formula KOH
CAS 1310-58-3
Packaging Poly Bottle

    For Laboratory,Research,or Manufacturing Use