missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Honeywell Fluka™ Potassium Hydroxide Solution, 1.0M, Honeywell Fluka™
Volumetric, 1.0 M KOH (1.0 N)
Manufacturer: Honeywell Fluka™ 31937625L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Potassium Hydroxide Solution | |
1.0M | |
KOH | |
MFCD00003553 | |
caswell no. 693, caustic potash, hydroxyde de potassium, potash lye, potassa, potasse caustique, potassium hydrate, potassium hydroxide, potassium hydroxide k oh, potassium hydroxide solution | |
[OH-].[K+] | |
56.105 | |
CHEBI:32035 | |
2.5L |
1310-58-3 | |
HKO | |
1.046g/mL at 25°C | |
1814 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium;hydroxide | |
14797 | |
56.11g/mol |
Chemical Identifiers
1310-58-3 | |
56.105 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium;hydroxide |
HKO | |
MFCD00003553 | |
caswell no. 693, caustic potash, hydroxyde de potassium, potash lye, potassa, potasse caustique, potassium hydrate, potassium hydroxide, potassium hydroxide k oh, potassium hydroxide solution | |
CHEBI:32035 | |
[OH-].[K+] |
Safety and Handling
P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H290-H314