missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium hydroxide solution, 0.1M in isopropanol, Honeywell™

Manufacturer:  Honeywell-Fluka 350081L

Catalog No. 6001240



Description

Description

Specifications

Specifications

Potassium hydroxide solution
0.1M in isopropanol
KOH
UN1219
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium;hydroxide
14797
56.11g/mol
1L
1310-58-3
HKO
MFCD00003553
caswell no. 693, caustic potash, hydroxyde de potassium, potash lye, potassa, potasse caustique, potassium hydrate, potassium hydroxide, potassium hydroxide k oh, potassium hydroxide solution
[OH-].[K+]
56.105
CHEBI:32035
12°C (53.6°F)

Chemical Identifiers

1310-58-3
56.105
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium;hydroxide
HKO
MFCD00003553
caswell no. 693, caustic potash, hydroxyde de potassium, potash lye, potassa, potasse caustique, potassium hydrate, potassium hydroxide, potassium hydroxide k oh, potassium hydroxide solution
CHEBI:32035
[OH-].[K+]
Safety and Handling

Safety and Handling

P210-P305 + P351 + P338-P370 + P378-P403 + P235

SDS
Documents