missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Hydroxide (Pellets/NF/FCC/EP/BP/JP), Fisher Chemical

$560.64 - $560.64

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.105
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium;hydroxide
SMILES [OH-].[K+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
P258212 SDS View Documents Thermo Scientific
P258212
2.5kg Poly Bottle Each for $560.64
 
Specifications

Chemical Identifiers

1310-58-3
56.105
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium;hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]

Specifications

Yellow
1310-58-3
2.0% max. (BP), 2.0% max. (BP/EP/JP), 3.5% max. (FCC), 2.0% max. (EP), 2.0% max. (JP), 3.5% max. (FCC)
Clear and Colorless (JP)
2.04g/cm³
Pass Test (BP), Pass Test (BP/EP/FCC/JP/NF), Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (NF)
0.003% max. (NF), 10ppm max. (BP/EP), 30ppm max. (JP), 10ppm max. (BP), 10ppm max. (EP), 30ppm max. (JP)
KWYUFKZDYYNOTN-UHFFFAOYSA-M
[OH-].[K+]
10ppm max. (BP), 10ppm max. (BP/EP), 10ppm max. (EP)
10mg/kg max. (FCC)
CHEBI:32035
Solid
0.1mg/kg max. (FCC)
Poly Bottle
3.5% max. (NF)
1.0% max. (BP), 1.0% max. (BP/EP), Pass Test (JP), 1.0% max. (EP), Pass Test (JP)
13.5
1320°C
2.5kg
Pass Test (BP), Pass Test (BP/EP), Pass Test (EP)
0.050% max. (JP), 50ppm max. (BP), 50ppm max. (BP/EP), 0.050% max. (JP), 50ppm max. (EP)
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
0.003% max. (NF), 0.003% max. (NF), 10ppm max. (BP/EP), 30ppm max. (JP)
Pass Test (FCC), Pass Test (FCC/NF), Pass Test (NF)
potassium;hydroxide
56.105
14797
56.11
85.0 to 100.5% (EP)
BP/EP/FCC/JP/NF
20ppm max. (BP), 20ppm max. (BP/EP), 20ppm max. (EP)
Meets Requirements (BP), Meets Requirements (BP/EP/NF), Meets Requirements (EP), Meets Requirements (NF)
50ppm max. (BP), 50ppm max. (BP/EP), 50ppm max. (EP)
360°C
Potassium Hydroxide
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive