Please login to your online account to display your discounted pricing

Potassium Hydroxide, Pellets, Multi-Compendial, N.F., J.T.Baker™

$226.83 - $557.36

Chemical Identifiers

Chemical Name or Material Potassium Hydroxide
CAS 1310-58-3
Purity Grade Multi-Compendial/NF
Molecular Formula KOH
Formula Weight 56.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003763

 
avantor performance
315201
Poly Bottle 500g Each for $226.83

02003771

 
avantor performance
315205
Poly Bottle 2.5kg Each for $557.36
Description & Specifications

Specifications

Purity Grade Multi-Compendial/NF
Physical Form Pellet
Formula Weight 56.11
Quantity 500g
Chemical Name or Material Potassium Hydroxide
Molecular Formula KOH
CAS 1310-58-3
Packaging Poly Bottle
Specific Gravity 1L = 2.04kg