Learn More
Potassium Hydroxide, in Isopropanol, Certified, 0.1000N ±0.0005N (0.1M), LabChem™
Supplier: LabChem LC195506
Specifications
Potassium Hydroxide | |
in Isopropanol | |
99.4,0.6 | |
KOH | |
0.1000N ±0.0005N (0.1M) | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
10 L | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
Certified | |
Cubitainer™ | |
Liquid |
Colorless | |
1310-58-3,67-63-0 | |
HKO | |
MFCD00003553 | |
UN1993 | |
Soluble in water | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
56.11 | |
Passes Test | |
Traceable to NIST |
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium hydroxide |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 | |
[OH-].[K+] |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
Danger
Recommended Storage : Room Temperature