missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Hydroxide, Certified, 10.0N ±0.1N (10M), LabChem™

Manufacturer:  LabChem LC193701

 View more versions of this product

Catalog No. LC193701



Specifications

Specifications

Potassium Hydroxide
500mL
59.69,40.31
1.39g/mL
KOH
Passes Test
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium;hydroxide
14797
56.11
Certified
Soluble in water
1.39g/mL
Colorless
1310-58-3,7732-18-5
10.0N ±0.1N (10M)
HKO
UN1814
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
[OH-].[K+]
56.105
CHEBI:32035
Liquid
Poly Bottle
Room Temperature
Traceable to NIST

Chemical Identifiers

1310-58-3
56.105
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
HKO
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium;hydroxide
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful to aquatic life.

GHS P Statement
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container in accordance with local, state, and federal regulations.

Danger

Documents
Promotions

Promotions