Please login to your online account to display your discounted pricing

Potassium Hydroxide, Certified, 0.0500N ±0.0005N (0.05M), LabChem™

$81.31 - $81.31

Chemical Identifiers

Chemical Name or Material Potassium Hydroxide
CAS 7732-18-5
CAS Max % 99.75
CAS 1310-58-3
CAS Max % 0.25
Purity Grade Certified Grade
Linear Formula KOH
Formula Weight 56.11
Synonym Caustic Potash
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC192802

 
lab chem
LC192802
Each for $81.31
Description & Specifications

Specifications

Chemical Name or Material Potassium Hydroxide
Color Clear, colorless
CAS 7732-18-5
CAS Max % 99.75
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 1310-58-3
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.25
Purity Grade Certified Grade
Concentration 0.0500N ±0.0005N (0.05M)
Physical Form Liquid
Density 1g/mL
Linear Formula KOH
Formula Weight 56.11
Identification Passes Test
Synonym Caustic Potash
Packaging Poly Bottle
Solubility Soluble in water
Storage Information Room Temperature
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.