missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium hydroxide, ca. 85%, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 424140025

 View more versions of this product

Catalog No. AC424140025



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium Hydroxide
85%
1310-58-3
ca. 85%
KOH
1813
15, 7761
2.5 kg
[OH-].[K+]
56.11
CHEBI:32035
≥85%
Pellet
White
1320.0°C
360.0°C
HKO
MFCD00003553
01,935; 02,346; 03,238; 04,409; 05,557; 06,486; 07,303; 08,415; 09,387; 11,439; 12,411; 13,258
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
56.11
ACS Reagent

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
Safety and Handling

Safety and Handling

Danger

EINECSNumber : 215-181-3

RTECSNumber : TT2100000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only