Please login to your online account to display your discounted pricing

Potassium Hydroxide, ACS Reagent Grade, Ricca Chemical


Quantity 500 gal.
Packaging Poly Wide mouth

Manufacturer: ricca chemical company  RIRDCP0500500B1

 View more versions of this product

Catalog No. RDCP0500500

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Deliquescent Pellets
Color White
Quantity 500 gal.
Chemical Name or Material Potassium Hydroxide
CAS 1310-58-3
CAS Min % 98.0
CAS Max % 100.0
Packaging Poly Wide mouth
CAS 7732-18-5 (Water, Distilled)

Specifications

Chemical Name or Material Potassium Hydroxide
CAS 1310-58-3
CAS Min % 98.0
CAS Max % 100.0
CAS 7732-18-5 (Water, Distilled)
Purity Grade ACS Reagent Grade
Physical Form Deliquescent Pellets