missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Hydroxide ACS AR Pellets, Macron Fine Chemicals™

macron

$302.60 - $302.60

Chemical Identifiers

CAS 1310-58-3
Physical Form Pellet
Molecular Formula KOH
Formula Weight 56.11
Chemical Name or Material Potassium Hydroxide
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK69841 Avantor Performance
698412
POLYSTORMOR 1kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK6984212 Avantor Performance
698406
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK6984500 Avantor Performance
698404
POLYSTORMOR 500g Each for $302.60
Case of 4 Each for $1,210.40
Specifications Description & Specifications

Chemical Identifiers

1310-58-3
KOH
Potassium Hydroxide
Pellet
56.11
ACS Grade

Specifications

1kg
Pellet
56.11
Potassium Hydroxide
1310-58-3
KOH
POLYSTORMOR
ACS Grade