Please login to your online account to display your discounted pricing

Potassium hydroxide, ACS, 85% min, K2CO3 2.0% max, Alfa Aesar™

In soap manufacture, in bleaching, as an electrolyte, as an absorbent, in dyestuffs, in liquid fertilizer, in electroplating, as a reagent

Chemical Identifiers

Chemical Name or Material Potassium hydroxide
CAS 1310-58-3
Purity Grade ACS Grade
Molecular Formula KOH
Formula Weight 56.11
MDL Number MFCD00003553
UN Number UN1813
Merck Index 14,7640
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1345118

 
alfa aesar
1345118
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Physical Form Pellet
Formula Weight 56.11
Quantity 50g
Odor Odorless
Assay ≥85%
Sensitivity Air Sensitive and Hygroscopic
Assay Percent Notes K2CO3 2.0% max
MDL Number MFCD00003553
UN Number UN1813
Merck Index 14,7640
Boiling Point 1320° to 1324°C
Melting Point 360°C
pH 13.5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H302;
Causes severe skin burns and eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POTASSIUM HYDROXIDE, SOLID
RTECS Number TT2100000
Chemical Name or Material Potassium hydroxide
TSCA TSCA
Molecular Formula KOH
CAS 1310-58-3
Solubility Information Soluble in water, alcohol, and glycerol. Slightly soluble in ether
Density 2.044
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H302;
Causes severe skin burns and eye damage.
Harmful if swallowed.