missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N

Manufacturer:  Alfa Aesar™ 88602LQ

 View more versions of this product

Catalog No. AA88602LQ



Specifications

Specifications

Potassium hydroxide Acculute Standard Volumetric Solution
1310-58-3
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
[OH-].[K+]
56.105
CHEBI:32035
Liquid
1 unit
Final Concentration 0.5N
HKO
UN1814
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium;hydroxide
14797
56.11
Colorless

Chemical Identifiers

1310-58-3
56.105
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium;hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POTASSIUM HYDROXIDE, SOLUTION

TSCA : TSCA

SDS Documents