Please login to your online account to display your discounted pricing

Potassium Hydroxide, 85.0% min., ACS, EMD Millipore

$159.31 - $1,046.09

Chemical Identifiers

Chemical Name or Material Potassium Hydroxide
Name Note For ACS analysis
CAS 1310-58-3
Physical Form Pellets
Assay Percent Range 85.0% min.
Molecular Formula KOH
Formula Weight 56.11
Synonym Caustic potash
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MPX14801

 
emd millipore corporation
PX14801
500g Each for $159.31

MPX14809

 
emd millipore corporation
PX14809
12kg Each for $1,046.09
Description & Specifications

Specifications

Physical Form Pellets
Formula Weight 56.11
Color White to off-white
Quantity 500g
Chemical Name or Material Potassium Hydroxide
Name Note For ACS analysis
Assay Percent Range 85.0% min.
Molecular Formula KOH
CAS 1310-58-3
Synonym Caustic potash