Please login to your online account to display your discounted pricing

Potassium Hydroxide, 85.0% min., ACS, EMD Millipore


Quantity 12kg

Manufacturer: emd millipore corporation  PX14809

 View more versions of this product

Catalog No. MPX14809

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Pellets
Formula Weight 56.11
Color White to off-white
Quantity 12kg
Packaging Poly Pail
CAS 1310-58-3
Molecular Formula KOH
Chemical Name or Material Potassium Hydroxide
Name Note For ACS analysis
Synonym Caustic potash
Assay Percent Range 85.0% min.

Specifications

Chemical Name or Material Potassium Hydroxide
Name Note For ACS analysis
CAS 1310-58-3
Physical Form Pellets
Assay Percent Range 85.0% min.
Molecular Formula KOH
Formula Weight 56.11
Synonym Caustic potash