Please login to your online account to display your discounted pricing

Potassium Hydroxide, 8.00 Normal, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Potassium Hydroxide
Name Note 8.00 Normal
CAS 1310-58-3
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 8.000 ± 0.005 N at 20°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R6255000100

 
ricca chemical company
RIR6255000100A
Poly natural 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R62550004A

 
ricca chemical company
RIR62550004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R6255000500

 
ricca chemical company
RIR6255000500A
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 100mL
Solvent or Matrix Aqueous Solution
Chemical Name or Material Potassium Hydroxide
CAS 1310-58-3
Solubility Solubility in Water: Infinite
Packaging Poly natural
Odor Odorless
Name Note 8.00 Normal
CAS 7732-18-5
Assay Percent Range 8.000 ± 0.005 N at 20°C