missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Hydroxide, 45% (w/w) Technical Grade, Ricca Chemical

Chemical Identifiers

CAS 1310-58-3,7732-18-5
CAS Min % 44.0,54.0
CAS Max % 46.0,56.0
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide
Grade Technical Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
6192-1 Ricca Chemical Company
RI61921
Poly natural 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
6192-32 Ricca Chemical Company
RI619232
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
6192-5 Ricca Chemical Company
RI61925
Cubitainer 20L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1310-58-3,7732-18-5
46.0,56.0
Potassium Hydroxide
44.0,54.0
Liquid
Technical Grade

Specifications

Liquid
4L
44.0,54.0
45% (w/w)
Potassium Hydroxide
Clear colorless
1310-58-3,7732-18-5
46.0,56.0
Poly natural
Technical Grade