Learn More
Potassium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 046330AP
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium hydroxide | |
Liquid | |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
25% w/v aq. soln. |
1310-58-3 | |
500 mL | |
KOH | |
UN1814 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium;hydroxide | |
14797 | |
56.11 |
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
[OH-].[K+] |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 |
Safety and Handling
GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POTASSIUM HYDROXIDE, SOLUTION
EINECSNumber : 215-181-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only