Please login to your online account to display your discounted pricing

Potassium Hydroxide, 2.00 Normal, Ricca Chemical

$95.58 - $95.58

Chemical Identifiers

Chemical Name or Material Potassium Hydroxide
CAS 7732-18-5
CAS Min % 78.0
CAS Max % 81.0
CAS 1310-58-3
CAS Min % 19.76
CAS Max % 20.57
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R62519001A

 
ricca chemical company
RIR62519001A
Poly natural 1L Each for $95.58

R625190020F

 
ricca chemical company
RIR625190020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R62519004A

 
ricca chemical company
RIR62519004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 1L
Concentration or Composition Notes 2.00Normal
Chemical Name or Material Potassium Hydroxide
CAS 7732-18-5
CAS Max % 81.0
CAS 1310-58-3
CAS Min % 19.76
CAS Min % 78.0
CAS Max % 20.57
Packaging Poly natural