missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 278400025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium hydroxide | |
1N solution in water | |
1310-58-3,7732-18-5 | |
KOH | |
1N, exact strength on the certificate of analysis | |
01,935; 02,346; 03,238; 04,409; 05,557; 06,486; 07,303; 08,415; 09,387; 11,439; 12,411; 13,258 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
2.5 L | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
Plastic bottle |
Colorless | |
100.0°C | |
HKO | |
MFCD00003553 | |
1814 | |
1.0500g/mL | |
Solubility in water: soluble. | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
56.11 | |
1.05 |
RUO – Research Use Only