missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Hydroxide, 1 N in Diethylene Glycol, Ricca Chemical

Supplier:  Ricca Chemical Company 62481

Catalog No. 62481

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Potassium Hydroxide
93.0,5.35
C4H10O3
diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol
MTHSVFCYNBDYFN-UHFFFAOYSA-N
2-(2-hydroxyethoxy)ethan-1-ol
8117
1 Normal in Diethylene Glycol
111-46-6,1310-58-3,7732-18-5
96.0,5.57
MFCD00002882
4 L
OCCOCCO
106.12
CHEBI:46807
Natural Poly Bottle

Chemical Identifiers

111-46-6
106.12
MTHSVFCYNBDYFN-UHFFFAOYSA-N
8117
2-(2-hydroxyethoxy)ethan-1-ol
C4H10O3
MFCD00002882
diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol
CHEBI:46807
OCCOCCO
Safety and Handling

Safety and Handling

ShelfLife : 12 months

SDS
Documents

Documents

Promotions

Promotions