missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Potassium Hydroxide, 1.00 N in Denatured Alcohol, Ricca Chemical
Chemical Identifiers
CAS |
1310-58-3 |
Molecular Formula |
HKO |
Molecular Weight (g/mol) |
56.11 |
InChI Key |
KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Synonym |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
PubChem CID |
14797 |
ChEBI |
CHEBI:32035 |
IUPAC Name |
potassium hydroxide |
SMILES |
[OH-].[K+] |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
1310-58-3 |
|
56.11 |
|
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
|
CHEBI:32035 |
|
[OH-].[K+] |
|
HKO |
|
KWYUFKZDYYNOTN-UHFFFAOYSA-M |
|
14797 |
|
potassium hydroxide |
Specifications
|
1310-58-3,7732-18-5 |
|
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
|
KWYUFKZDYYNOTN-UHFFFAOYSA-M |
|
potassium hydroxide |
|
14797 |
|
1.00N in Alcohol, in SDA-3A (100 parts Ethanol, 5 parts Methanol) |
|
Potassium Hydroxide |
|
HKO |
|
1 L |
|
[OH-].[K+] |
|
56.11 |
|
CHEBI:32035 |
|
Natural Poly Bottle |
SDS
missing translation for 'documents'