missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Hydroxide, 1.00 N in Denatured Alcohol, Ricca Chemical
Supplier: Ricca Chemical Company 626691
Specifications
Potassium Hydroxide | |
HKO | |
4 L | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
Natural Poly Bottle |
1310-58-3,7732-18-5 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
1.00N in Alcohol, in SDA-3A (100 parts Ethanol, 5 parts Methanol) |
Chemical Identifiers
1310-58-3 | |
56.11 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 | |
[OH-].[K+] |
HKO | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium hydroxide |