Please login to your online account to display your discounted pricing

Potassium Hydroxide, 1.00N in Methanol, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Potassium Hydroxide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

6262-1

 
ricca chemical company
RI62621
Plastic 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

6262-32

 
ricca chemical company
RI626232
Plastic 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R62620004F

 
ricca chemical company
RIR62620004F
Cubitainer 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 4L
Chemical Name or Material Potassium Hydroxide
Concentration or Composition (by Analyte or Components) 1.00N in Methanol
Packaging Plastic