missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Hydroxide, 1.00 Normal, Ricca Chemical

Manufacturer:  Ricca Chemical Company RIR62500004F

 View more versions of this product

Catalog No. R62500004F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Potassium Hydroxide
HKO
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium;hydroxide
14797
Liquid
1.00N
4L
1310-58-3,7732-18-5
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
[OH-].[K+]
56.105
CHEBI:32035
Cubitainer
Colorless

Chemical Identifiers

1310-58-3
56.105
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
HKO
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium;hydroxide
Documents