missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Hydroxide, 0.200 N (N/5), Ricca Chemical
Supplier: Ricca Chemical Company 621032
Specifications
Potassium Hydroxide | |
HKO | |
1 L | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
Natural Poly Bottle |
1310-58-3,7732-18-5 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
0.200N (N/5) |
Chemical Identifiers
1310-58-3 | |
56.11 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 | |
[OH-].[K+] |
HKO | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium hydroxide |