Please login to your online account to display your discounted pricing

Potassium Hydroxide, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$212.46 - $662.34

Chemical Identifiers

Chemical Name or Material Potassium Hydroxide
CAS 1310-58-3
Name Note 0.1N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula KOH
Formula Weight 56.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003685

 
avantor performance
560303
Cubetainer 4L Each for $662.34

02003684

 
avantor performance
560302
Narrow Mouth Polyethylene Bottle 1L Each for $212.46
Description & Specifications

Specifications

Formula Weight 56.11
Quantity 4L
Chemical Name or Material Potassium Hydroxide
Name Note 0.1N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula KOH
CAS 1310-58-3
Packaging Cubetainer
Specific Gravity 1L = 1.05kg