missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium hydroxide, 0.1N Standardized Solution, Alfa Aesar™

$32.78 - $175.47

Chemical Identifiers

CAS 1310-58-3
Physical Form Liquid
Molecular Formula HKO
Formula Weight 56.11
MDL Number MFCD00003553
Chemical Name or Material Potassium hydroxide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA35599K2 SDS Alfa Aesar
35599K2
1L Each for $32.78
AA35599K7 SDS Alfa Aesar
35599K7
4L Each for $73.89
AA35599M4 SDS Alfa Aesar
35599M4
20L Each for $175.47
Specifications Description & Specifications

Chemical Identifiers

1310-58-3
HKO
MFCD00003553
Liquid
56.11
Potassium hydroxide

Specifications

56.11
Colorless
1310-58-3
Colorless liquid
23 hPa (17mm Hg) at 20°C
1L
Liquid
MFCD00003553
HKO
Potassium hydroxide

Safety and Handling

H315-H319: Causes skin irritation. Causes serious eye irritation.

EINECSNumber : 215-181-3