Please login to your online account to display your discounted pricing

Potassium Hydroxide, 0.1N in Methanol Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$125.77 - $855.26

Chemical Identifiers

Chemical Name or Material Potassium Hydroxide
CAS 1310-58-3
Name Note 0.1N in Methanol Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula KOH
Formula Weight 56.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003680

 
avantor performance
565002
Narrow Mouth Polyethylene Bottle 1L Each for $125.77

02003681

 
avantor performance
565003
Cubetainer 4L Each for $855.26
Description & Specifications

Specifications

Formula Weight 56.11
Quantity 1L
Chemical Name or Material Potassium Hydroxide
Molecular Formula KOH
CAS 1310-58-3
Name Note 0.1N in Methanol Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 0.80kg
Flash Point 12.00°C