Please login to your online account to display your discounted pricing

Potassium Hydroxide, 0.100 Normal, Ricca Chemical


Quantity 4L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR62000004F

 View more versions of this product

Catalog No. R62000004F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Potassium Hydroxide
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 1310-58-3
CAS Min % 0.63
CAS Max % 0.65
Packaging Cubitainer
Concentration or Composition Notes 0.100N
Quantity 4L

Specifications

Chemical Name or Material Potassium Hydroxide
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 1310-58-3
CAS Min % 0.63
CAS Max % 0.65