missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium hydrotris(1-pyrazolyl)borate, Thermo Scientific Chemicals
$126.45 - $467.11
Chemical Identifiers
CAS | 18583-60-3 |
---|---|
Molecular Formula | C9H10BKN6 |
Molecular Weight (g/mol) | 252.13 |
MDL Number | MFCD00040346 |
InChI Key | IBNGZWIUPDMZMG-UHFFFAOYSA-N |
Synonym | potassium hydrotris 1-pyrazolyl borate, potassium hydridotri 1h-pyrazol-1-yl borate 1-, potassium hydro-tris 1-pyrazolyl borate, potassium tri 1h-pyrazol-1-yl hydroborate, potassium tri 1-pyrazolyl borohydride, potassium hydrotris 1-pyrazolato borate, hydrate |
IUPAC Name | potassium tris(1H-pyrazol-1-yl)boranuide |
SMILES | [K+].C1=CN(N=C1)[BH-](N1C=CC=N1)N1C=CC=N1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18583-60-3 | |
252.13 | |
IBNGZWIUPDMZMG-UHFFFAOYSA-N | |
potassium tris(1H-pyrazol-1-yl)boranuide |
C9H10BKN6 | |
MFCD00040346 | |
potassium hydrotris 1-pyrazolyl borate, potassium hydridotri 1h-pyrazol-1-yl borate 1-, potassium hydro-tris 1-pyrazolyl borate, potassium tri 1h-pyrazol-1-yl hydroborate, potassium tri 1-pyrazolyl borohydride, potassium hydrotris 1-pyrazolato borate, hydrate | |
[K+].C1=CN(N=C1)[BH-](N1C=CC=N1)N1C=CC=N1 |
Specifications
18583-60-3 | |
C9H10BKN6 | |
1 g | |
IBNGZWIUPDMZMG-UHFFFAOYSA-N | |
potassium tris(1H-pyrazol-1-yl)boranuide | |
252.13 | |
Potassium hydrotris(1-pyrazolyl)borate |
185°C to 189°C | |
MFCD00040346 | |
potassium hydrotris 1-pyrazolyl borate, potassium hydridotri 1h-pyrazol-1-yl borate 1-, potassium hydro-tris 1-pyrazolyl borate, potassium tri 1h-pyrazol-1-yl hydroborate, potassium tri 1-pyrazolyl borohydride, potassium hydrotris 1-pyrazolato borate, hydrate | |
[K+].C1=CN(N=C1)[BH-](N1C=CC=N1)N1C=CC=N1 | |
252.13 | |
Crystalline |
Safety and Handling
P231+P232-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H261-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only