missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Hydrogen Sulfate ACS AR Powder, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7646-93-7
Physical Form Powder
Molecular Formula KHSO4
Formula Weight 136.17
Chemical Name or Material Potassium Hydrogen Sulfate, Fused
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK7112500 Avantor Performance
711204
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7646-93-7
KHSO4
Potassium Hydrogen Sulfate, Fused
Powder
136.17
ACS Grade

Specifications

136.17
7646-93-7
POLYSTORMOR
Potassium Hydrogen Sulfate, Fused
Powder
KHSO4
500g
ACS Grade