missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium hydrogen sulfate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1242836

 View more versions of this product

Catalog No. AAA1242836



Description

Description

Potassium hydrogen sulfate is used in the conversion of tartrates to bitartrates in wine. It serves as a disintegrating agent in analytical chemistry and as an intermediate in the preparation of potassium persulfate by electrolysis. It is also employed in the manufacturing of fertilizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen sulfate is used in the conversion of tartrates to bitartrates in wine. It serves as a disintegrating agent in analytical chemistry and as an intermediate in the preparation of potassium persulfate by electrolysis. It is also employed in the manufacturing of fertilizers.

Solubility
Soluble in water, acetone and ethanol.

Notes
Hygroscopic. Incompatible with strong bases, strong oxidizing agents and alcohols.
Specifications

Specifications

Potassium hydrogen sulfate
7646-93-7
HKO4S
MFCD00011404
14,7613
Soluble in water,acetone and ethanol.
[K+].OS([O-])(=O)=O
136.16
136.17
Odorless
2.24
195°C to 197°C (decomposition)
500 g
KHSO4
UN2509
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
CHKVPAROMQMJNQ-UHFFFAOYSA-M
potassium;hydrogen sulfate
516920
97%
Hygroscopic

Chemical Identifiers

7646-93-7
136.16
CHKVPAROMQMJNQ-UHFFFAOYSA-M
516920
[K+].OS([O-])(=O)=O
HKO4S
MFCD00011404
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
potassium;hydrogen sulfate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POTASSIUM HYDROGEN SULFATE

EINECSNumber : 231-594-1

RTECSNumber : TS7200000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only