missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium hydrogen phosphate, ACS, 98.0% min., Alfa Aesar™

Manufacturer:  Alfa Aesar 1159330

 View more versions of this product

Catalog No. AA1159330



Specifications

Specifications

Potassium hydrogen phosphate
98.0% min.
MFCD00011383
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
OP(=O)([O-])[O-].[K+].[K+]
174.17
CHEBI:32031
Powder
Maximum level of impurities: Insoluble matter 0.01%, Loss on drying at 105 1.0%, pH of a 5% solution 8.5 to 9.6 at 25° , Cl 0.003%, Nitrogen compounds (as N) 0.001%, SO4 0.005%, Heavy Metals (as Pb)
250g
7758-11-4
HK2O4P
White powder
ZPWVASYFFYYZEW-UHFFFAOYSA-L
dipotassium;hydrogen phosphate
24450
174.18
ACS
White

Chemical Identifiers

7758-11-4
174.17
ZPWVASYFFYYZEW-UHFFFAOYSA-L
24450
dipotassium;hydrogen phosphate
HK2O4P
MFCD00011383
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
CHEBI:32031
OP(=O)([O-])[O-].[K+].[K+]
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. This material and/or its container must be disposed of as hazardous waste.

H319: Causes serious eye irritation.

EINECSNumber : 231-834-5

Documents