Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hydrogen iodate, 0.025N Standardized Solution

Chemical Identifiers

Chemical Name or Material Potassium hydrogen iodate
CAS 13455-24-8
Name Note Standardized Solution
Formula Weight 389.91
MDL Number MFCD00011400
Synonym Potassium biiodate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35600K2

 
alfa aesar™
35600K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 389.91
Chemical Name or Material Potassium hydrogen iodate
TSCA TSCA
Quantity 1L
Name Note Standardized Solution
Physical Form Liquid
Concentration 0.025N
MDL Number MFCD00011400
CAS 13455-24-8
Synonym Potassium biiodate
Odor Odorless