Learn More
Potassium hydrogen fluoride, 99+% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 011557A3
Description
Potassium hydrogen fluoride is widely used as an etchant in glass and cleaning products. It finds an important application in the synthesis of special optical glass viz. crown and crown flint glass and as a catalyst for polymerization. It is involved in the manufacturing of wood preservatives, soldering agents and brazing. It is also used in the manufacturing of organic and inorganic fluorine compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Electrolyte, flux, catalyst in alkylation of benzene with olefins, and in the synthesis of pure KF
Specifications
Potassium hydrogen fluoride | |
7789-29-9 | |
Crystalline | |
(metals basis) | |
UN1811 | |
Potassium bifluoride | |
78.1 | |
Odorless | |
2.370 g/mL |
239°C (decomposition) | |
1 | |
2 kg | |
MFCD00011403 | |
14,7610 | |
Soluble in water and ethanol. | |
≥99% | |
Hygroscopic |
Chemical Identifiers
7789-29-9 | |
MFCD00011403 | |
Potassium bifluoride |
Safety and Handling
GHS H Statement
H301-H314
Toxic if swallowed.
Causes severe skin burns and eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POTASSIUM HYDROGENDIFLUORIDE SOLID
EINECSNumber : 232-156-2
RTECSNumber : TS6650000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only