missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium hydrogen fluoride, ≥99%, extra pure, Acros Organics

Manufacturer:  Thermo Fisher Scientific 219235000

 View more versions of this product

Catalog No. AC219235000


 

Specifications Description & Specifications

Specifications

Extra Pure
≥99.0%
500g
16,283
1811
99+%
7789-29-9
KHF2
Potassium hydrogen fluoride
50ppm max.
50ppm max.
Crystals or Crystalline Powder
78.1
White to light gray
15, 7731
Potassium bifluoride
500ppm max.
99+%
HF2K
MFCD00011403
Plastic bottle

Chemical Identifiers

7789-29-9
HF2K
78.1
Potassium bifluoride
≥99.0%
99+%
KHF2
MFCD00011403
Potassium hydrogen fluoride, 99+%
Extra Pure

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 232-156-2

RTECSNumber : TS6650000

TSCA : TSCA