Please login to your online account to display your discounted pricing

Potassium hydrogen diiodate, 99.85%, extra pure, Acros Organics

$178.19 - $625.12

Chemical Identifiers

Chemical Name or Material Potassium hydrogen diiodate
Name Note p.a.
CAS 13455-24-8
Purity Grade Extra Pure
Assay ≥99.80%
Assay Percent Range 99.85%
Molecular Formula HI2KO6
Linear Formula KH(IO3)2
Formula Weight 389.9
MDL Number MFCD00011400
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC211471000

  SDS
thermo fisher scientific
211471000
Glass bottle 100g Each for $178.19

AC211475000

  SDS
thermo fisher scientific
211475000
Glass bottle 500g Each for $625.12
Description & Specifications

Specifications

Assay ≥99.80%
Formula Weight 389.9
Purity Grade Extra Pure
Physical Form Crystals or Powder
Quantity 100g
Color White to slightly yellow
TSCA TSCA
EINECS Number 236-650-9
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.
Molecular Formula HI2KO6
Linear Formula KH(IO3)2
MDL Number MFCD00011400
CAS 13455-24-8
Assay Percent Range 99.85%
Chemical Name or Material Potassium hydrogen diiodate
Name Note p.a.
Insoluble Matter 0.01% max.
Additional Information chloride, chlorate, bromide, bromate (as Cl): < 0.02%
Iron (Fe) 10ppm max.
Packaging Glass bottle
Sulfate 50ppm max.
Health Hazard 1 GHS Signal Word: Danger

Oxidation reagent

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.