missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hydrogen carbonate, 99%

Manufacturer:  Alfa Aesar™ A124290E

 View more versions of this product

Catalog No. AAA124290E


 

Description

Description

Applications
Potassium hydrogen carbonate is used in foods to regulate pH, as a fire suppression agent (BC dry chemical) and as a buffering agent in medications. It acts as an additive in winemaking and is an active component in club soda. Furthermore, it finds use in crops for the neutralization of acidic soils. It is also used as a source of carbon dioxide for leavening in baking. It is utilized to inhibit the growth of aspergillus parasiticus in czapek′s agar. It is employed as a catalyst in synthetic fiber polymerization and olefin dehydrogenation.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

Potassium hydrogen carbonate
99%
MFCD00011402
Potassium bicarbonate
C(=O)(O)[O-].[K+]
100.114
CHEBI:81862
Odorless
298-14-6
CHKO3
14,7609
TYJJADVDDVDEDZ-UHFFFAOYSA-M
potassium;hydrogen carbonate
516893
100.12
2500g

Chemical Identifiers

298-14-6
100.114
TYJJADVDDVDEDZ-UHFFFAOYSA-M
516893
CHKO3
MFCD00011402
Potassium bicarbonate
CHEBI:81862
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents