missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hydride, 30% w/w in mineral oil

Manufacturer:  Alfa Aesar™ L1326622

 View more versions of this product

Catalog No. AAL1326622



Description

Description

Applications
Potassium hydride is a precursor in the production of various bases by deprotonation, highly-hindered bases potassium triethylmethoxide and potassium 3-aminopropylamide It serves as a powerful base in organic synthesis of deprotonate carbonyl compounds and amines to form enolates and amides respectively.

Solubility
Insoluble in benzene, diethyl ether and carbon disulfide.

Notes
Moisture sensitive. Incompatible and reacts violently with water, acids, alcohols, oxidizing agents and chlorinated solvents.
Specifications

Specifications

Potassium hydride
HK
UN1409
OCFVSFVLVRNXFJ-UHFFFAOYSA-N
potassium;hydride
82127
30% w/w in mineral oil
100g
7693-26-7
MFCD00011357
potassium hydride, potassium hydride kh, kaliumhydrid, acmc-20ajuo, kh, potassium hydride w/w in mineral oil
[H-].[K+]
40.106
40.11
Odorless
160°C (320°F)

Chemical Identifiers

7693-26-7
40.106
OCFVSFVLVRNXFJ-UHFFFAOYSA-N
82127
[H-].[K+]
HK
MFCD00011357
potassium hydride, potassium hydride kh, kaliumhydrid, acmc-20ajuo, kh, potassium hydride w/w in mineral oil
potassium;hydride
Safety and Handling

Safety and Handling

GHS H Statement
H260-H314
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.

GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: METAL HYDRIDES, WATER REACTIVE, N.O.S.

TSCA : TSCA

Documents